missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 15 Mouse anti-Human, Clone: KRT15/2958, Novus Biologicals™
Mouse Monoclonal Antibody
£215.00
Specifications
| Antigen | Cytokeratin 15 |
|---|---|
| Clone | KRT15/2958 |
| Concentration | 0.2 mg/ml |
| Dilution | Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin), Peptide Array |
Description
Cytokeratin 15 Monoclonal specifically detects Cytokeratin 15 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry-Paraffin, Protein Array.Specifications
| Cytokeratin 15 | |
| 0.2 mg/ml | |
| Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin), Peptide Array | |
| Unconjugated | |
| Mouse | |
| Cell Biology, Cellular Markers | |
| 10 mM PBS with 0.05% BSA | |
| 3866 | |
| IgG2b κ | |
| Protein A or G purified |
| KRT15/2958 | |
| Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CK15, CK-15, cytokeratin 15, cytokeratin-15, K15keratin-15, basic, K1CO, keratin 15, keratin, type I cytoskeletal 15, Keratin-15, keratin-15, beta, KRTB, type I cytoskeletal 15 | |
| Recombinant full-length human Cytokeratin 15 protein | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title