missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 15 Antibody (SPM190), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-53313-20UG
This item is not returnable.
View return policy
Description
Cytokeratin 15 Monoclonal specifically detects Cytokeratin 15 in Human, Rat, Bovine samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| Cytokeratin 15 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Western Blot 0.5 - 1 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunohistochemistry-Paraffin 0.5 - 1 ug/ml, Immunohistochemistry-Frozen 0.5 - 1.0 ug/ml | |
| CK15, CK-15, cytokeratin 15, cytokeratin-15, K15keratin-15, basic, K1CO, keratin 15, keratin, type I cytoskeletal 15, Keratin-15, keratin-15, beta, KRTB, type I cytoskeletal 15 | |
| Mouse | |
| 52 kDa | |
| 20 μg | |
| Cytoskeleton Markers | |
| 3866 | |
| Store at 4C. | |
| IgG2a κ |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| SPM190 | |
| Unconjugated | |
| 10mM PBS with 0.05% BSA with 0.05% Sodium Azide | |
| KRT15 | |
| A 17 amino acids synthetic peptide from the C- terminal of Human Cytokeratin 15 | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Rat, Bovine | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction