missing translation for 'onlineSavingsMsg'
Learn More

Cytokeratin 15 Antibody (SPM190), DyLight 680, Novus Biologicals™

Product Code. 18418114 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18418114 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18418114 Supplier Novus Biologicals Supplier No. NBP254453FR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Cytokeratin 15 Monoclonal antibody specifically detects Cytokeratin 15 in Human, Rat, Bovine samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), CyTOF
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytokeratin 15
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), CyTOF
Classification Monoclonal
Clone SPM190
Conjugate DyLight 680
Formulation 50 mM Sodium Borate
Gene Alias CK15, CK-15, cytokeratin 15, cytokeratin-15, K15keratin-15, basic, K1CO, keratin 15, keratin, type I cytoskeletal 15, Keratin-15, keratin-15, beta, KRTB, type I cytoskeletal 15
Host Species Mouse
Immunogen A 17 amino acids synthetic peptide from the C- terminal of Human Cytokeratin 15 (Uniprot: P19012)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers
Primary or Secondary Primary
Gene ID (Entrez) 3866
Target Species Human, Rat, Bovine
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.