missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cytokeratin 14 Recombinant Rabbit Monoclonal Antibody (5Z4O9)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA535220
This item is not returnable.
View return policy
Description
Immunogen sequence: VGGDVNVEMD AAPGVDLSRI LNEMRDQYEK MAEKNRKDAE EWFFTKTEEL NREVATNSEL VQSGKSEISE LRRTMQNLEI ELQSQLSMKA SLENSLEETK GRYCMQLAQI QEMIGSVEEQ LAQLRCEMEQ QNQEYKILLD VKTRLEQEIA TYRRLLEGED AHLSSSQFSS GSQSSRDVTS SSRQIRTKVM DVHD.
Cytokeratins (CK) are intermediate filaments of epithelial cells, both in keratinizing tissue (ie. skin) and non-keratinizing cells (ie. mesothelial cells). At least 20 different CKs can be identified. Biochemically, most members of the CK family fall into one of two classes, type I (acidic polypeptides) and type II (basic polypeptides). Belonging to the type I subfamily of low molecular weight keratins and existing in combination with keratin 5, keratin 14 distinguishes stratified epithelial cells from simple epithelial cells and is useful in identification of squamous cell carcinomas. It is considered a prognostic marker in breast carcinomas. At least one member of the acidic family and one member of the basic family is expressed in all epithelial cells.
Specifications
| Cytokeratin 14 | |
| Recombinant Monoclonal | |
| 1.25 mg/mL | |
| PBS with 50% glycerol, 0.05% BSA and 0.02% sodium azide; pH 7.3 | |
| P02533, Q61781, Q6IFV1 | |
| KRT14 | |
| Recombinant protein (or fragment). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 5Z4O9 | |
| Unconjugated | |
| KRT14 | |
| AI626930; CK14; CK-14; cytokeratin 14; cytokeratin-14; EBS3; EBS4; epidermolysis bullosa simplex, Dowling-Meara, Koebner; K14; Ka14; keratin 14; keratin 14 (epidermolysis bullosa simplex, Dowling-Meara, Koebner); keratin 14, type I; keratin complex 1, acidic, gene 14; keratin, type I cytoskeletal 14; keratin, type I cytoskeletal 17; keratin-14; Koebner; Krt-1.14; Krt1-14; KRT14; LOC100737113; NFJ; Type I keratin Ka14 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 16664, 287701, 3861 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto