missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 14 Mouse anti-Human, Mouse, Rat, Porcine, Clone: KRT14/2375, Novus Biologicals™
Mouse Monoclonal Antibody
£396.00
Specifications
| Antigen | Cytokeratin 14 |
|---|---|
| Clone | KRT14/2375 |
| Concentration | 0.2 mg/mL |
| Dilution | Flow Cytometry 1 to 2 μg/million cells, Immunohistochemistry-Paraffin 1 to 2 μg/mL |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin) |
Description
Cytokeratin 14 Monoclonal specifically detects Cytokeratin 14 in Human, Mouse, Rat, Porcine samples. It is validated for Flow Cytometry, Immunohistochemistry-Paraffin.Specifications
| Cytokeratin 14 | |
| 0.2 mg/mL | |
| Flow Cytometry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Cell Biology, Cellular Markers | |
| 10 mM PBS with 0.05% BSA | |
| 3861 | |
| IgG2a κ | |
| Protein A or G purified |
| KRT14/2375 | |
| Flow Cytometry 1 to 2 μg/million cells, Immunohistochemistry-Paraffin 1 to 2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat, Pig | |
| CK14, CK-14, cytokeratin 14, cytokeratin-14, EBS3, EBS4, K14, keratin 14, keratin 14 (epidermolysis bullosa simplex, Dowling-Meara, Koebner), keratin, type I cytoskeletal 14, Keratin-14, NFJ | |
| Recombinant human Cytokeratin 14 fragment (around aa351-472) (exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title