missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 14 Antibody (KRT14/532), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-47720R
This item is not returnable.
View return policy
Description
Cytokeratin 14 Monoclonal antibody specifically detects Cytokeratin 14 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunofluorescence, CyTOF
Specifications
| Cytokeratin 14 | |
| Monoclonal | |
| DyLight 550 | |
| CK14, CK-14, cytokeratin 14, cytokeratin-14, EBS3, EBS4, K14, keratin 14, keratin 14 (epidermolysis bullosa simplex, Dowling-Meara, Koebner), keratin, type I cytoskeletal 14, Keratin-14, NFJ | |
| Recombinant full-length human Cytokeratin 14 protein (Uniprot: P02533) | |
| 0.1 mL | |
| Cell Biology, Cellular Markers | |
| 3861 | |
| Store at 4°C in the dark. | |
| IgG3 κ |
| Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunofluorescence | |
| KRT14/532 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction