missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 14 Antibody (CL14565) [CoraFluor™ 1], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-26868CL1
This item is not returnable.
View return policy
Description
Cytokeratin 14 Monoclonal antibody specifically detects Cytokeratin 14 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
| Cytokeratin 14 | |
| Monoclonal | |
| CoraFluor 1 | |
| CK14, CK-14, cytokeratin 14, cytokeratin-14, EBS3, EBS4, K14, keratin 14, keratin 14 (epidermolysis bullosa simplex, Dowling-Meara, Koebner), keratin, type I cytoskeletal 14, Keratin-14, NFJ | |
| This antibody was developed using a synthetic peptide derived from P02533, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Cell Biology, Cellular Markers | |
| 3861 | |
| Store at 4⪚C in the dark. Do not freeze. | |
| IgG2b |
| Immunohistochemistry, Western Blot, Immunocytochemistry/Immunofluorescence | |
| CL14565 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction