Learn More
Invitrogen™ Cytokeratin 13 Monoclonal Antibody (3G4)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA517109
Description
MA5-17109 targets KRT13 in FACS, indirect ELISA, and WB applications and shows reactivity with Human samples. The MA5-17109 immunogen is purified recombinant fragment of human KRT13 (amino acids: 143-295) expressed in E. Coli. MA5-17109 detects KRT13 which has a predicted molecular weight of approximately 52kDa.
Cytokeratins are a subfamily of intermediate filament proteins and are characterized by a remarkable biochemical diversity, represented in human epithelial tissues by at least 20 different polypeptides. They range in molecular weight between ~40 kDa and ~68 kDa and isoelectric pH between 4.9 - 7.8. The individual human cytokeratins are numbered 1 to 20. The various epithelia in the human body usually express cytokeratins which are not only characteristic of the type of epithelium, but also related to the degree of maturation or differentiation within an epithelium. Cytokeratin subtype expression patterns are used to an increasing extent in the distinction of different types of epithelial malignancies. The cytokeratin antibodies are not only of assistance in the differential diagnosis of tumors using immunohistochemistry on tissue sections, but are also a useful tool in cytopathology and flow cytometric assays.
Specifications
| Cytokeratin 13 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide | |
| P13646 | |
| KRT13 | |
| Purified recombinant fragment of human KRT13 (amino acids: 143-295) expressed in E. Coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Western Blot | |
| 3G4 | |
| Unconjugated | |
| KRT13 | |
| 47 kDa cytokeratin; CK13; CK-13; cytokeratin 13; cytokeratin-13; K13; K-13; Ka13; keratin 13; keratin 13, type I; keratin complex 1, acidic, gene 13; keratin, type I cytoskeletal 13; Keratin-13; Krt-1.13; Krt1-13; KRT13; krt13 {ECO:0000250; MGC161462; MGC3781; Type I keratin Ka13; UniProtKB:P13646}; WSN2 | |
| Mouse | |
| Protein G | |
| RUO | |
| 3860 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.