missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 12 Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
£180.00 - £409.00
Specifications
| Antigen | Cytokeratin 12 |
|---|---|
| Dilution | Western Blot 1:100 - 1:500, ELISA Recommended starting concentration is 1 μg/mL, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Applications | ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30232443
|
Novus Biologicals
NBP3-33197-100ul |
100 μL |
£409.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30229204
|
Novus Biologicals
NBP3-33197-20ul |
20 μL |
£180.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cytokeratin 12 Monoclonal antibody specifically detects Cytokeratin 12 in Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ImmunofluorescenceSpecifications
| Cytokeratin 12 | |
| ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Cytoskeleton Markers, Neuroscience, Signal Transduction | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| 3859 | |
| IgG | |
| Affinity purified |
| Western Blot 1:100 - 1:500, ELISA Recommended starting concentration is 1 μg/mL, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mouse, Rat | |
| keratin, type I cytoskeletal 12, CK12, K12, keratin 12 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 250-494 of human Cytokeratin 12 (KRT12) (NP_000214.1).,, Sequence:, EMQIESLNEELAYMKKNHEDELQSFRVGGPGEVSVEMDAAPGVDLTRLLNDMRAQYETIAEQNRKDAEAWFIEKSGELRKEISTNTEQLQSSKSEVTDLRRAFQNLEIELQSQLAMKKSLEDSLAEAEGDYCAQLSQVQQLISNLEAQLLQVRADAERQNVDHQRLLNVKARLELEIETYRRLLDGEAQGDGLEESLFVTDSKSQAQSTDSSKDPTKTRKIKTVVQEMVNGEVVSSQVQEIEELM | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title