missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 10 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18343-1-AP-150UL
This item is not returnable.
View return policy
Description
Cytokeratins (CK) are intermediate filaments of epithelial cells, both in keratinizing tissue (i.e.., skin) and non-keratinizing cells (i.e.. mesothelial cells). Although not a traditional marker for endothelial cells, cytokeratins have also been found in some microvascular endothelial cells. At least 20 different cytokeratins (CK) in the molecular range of 40-70 kDa and isoelectric points of 5-8.5 can be identified using two dimensional gel electrophoresis. Biochemically, most members of the CK family fall into one of two classes, type I (acidic polypeptides) and type II (basic polypeptides). At least one member of the acidic family and one member of the basic family is expressed in all epithelial cells. Monoclonal antibodies to cytokeratin proteins can be useful markers for tumor identification and classification.Specifications
| Cytokeratin 10 | |
| Polyclonal | |
| Unconjugated | |
| KRT10 | |
| CK 10, CK10, Cytokeratin 10, K10, keratin 10, KPP, KRT10 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 16661, 3858, 450225 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin) | |
| 0.24 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P02535, P13645, Q6IFW6 | |
| Krt10 | |
| Cytokeratin 10 Fusion Protein Ag13136 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction