missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cytokeratin 10 Monoclonal Antibody (DE-K10)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA513705
This item is not returnable.
View return policy
Description
MA5-13705 targets Cytokeratin 10 in IF, IHC (P), and WB applications and shows reactivity with Canine, Feline, Human, and Rat samples. The MA5-13705 immunogen is cytoskeletal preparation extracted from human ectocervical epithelium.
Cytokeratins (CK) are intermediate filaments of epithelial cells, both in keratinizing tissue (i.e., skin) and non-keratinizing cells (i.e. mesothelial cells). Although not a traditional marker for endothelial cells, cytokeratins have also been found in some microvascular endothelial cells. At least 20 different cytokeratins (CK) in the molecular range of 40-70 kDa and isoelectric points of 5-8. 5 can be identified using two dimensional gel electrophoresis. Biochemically, most members of the CK family fall into one of two classes, type I (acidic polypeptides) and type II (basic polypeptides). At least one member of the acidic family and one member of the basic family is expressed in all epithelial cells. Monoclonal antibodies to cytokeratin proteins can be useful markers for tumor identification and classification.
Specifications
| Cytokeratin 10 | |
| Monoclonal | |
| 0.2 mg/mL | |
| PBS with 0.05% BSA and 0.05% sodium azide; pH 7.4 | |
| P13645, Q6EIZ0, Q6IFW6 | |
| KRT10 | |
| Cytoskeletal preparation extracted from human ectocervical epithelium. | |
| 500 μL | |
| Primary | |
| Canine, Feline, Human, Rat | |
| Antibody | |
| IgG1 κ |
| Immunohistochemistry (Paraffin), Western Blot | |
| DE-K10 | |
| Unconjugated | |
| KRT10 | |
| 56 kDa cytokeratin; BCIE; BIE; CK10; CK-10; CK-17; cytokeratin 10; cytokeratin-10; cytokeratin-17; D130054E02Rik; EHK; epidermal keratin 10; epithelial keratin 10; intermediate filament protein; K10; K1C1; Ka10; Ker10; keratin 10; keratin 10 (epidermolytic hyperkeratosis; keratosis palmaris et plantaris); keratin 10, type I; keratin complex 1, acidic, gene 10; keratin, type I cytoskeletal 10; Keratin, type I cytoskeletal 59 kDa; keratin-10; keratin-17; KPP; Krt-1.10; KRT10; krt10 {ECO:0000250; Krt1-10; suprabasal cytokeratin 10; type I keratin KA10; UniProtKB:P13645} | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 101085587, 3858, 450225, 491006 | |
| 4°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction