missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 10/13 Antibody (DE-K13), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-45256-0.1MG
This item is not returnable.
View return policy
Description
Ensure accurate, reproducible results in Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence
Cytokeratin 10/13 Monoclonal specifically detects Cytokeratin 10/13 in Human, Feline samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunofluorescence.
Specifications
| Cytokeratin 10/13 | |
| Monoclonal | |
| 0.2mg/mL | |
| Western Blot 0.25 - 0.5 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, SDS-Page, Immunofluorescence 0.5 - 1.0 ug/ml | |
| BCIE, BIE, CK10, CK-10, cytokeratin 10, Cytokeratin-10, EHK, K10keratosis palmaris et plantaris, keratin 10, keratin, type I cytoskeletal 10, keratin-10, KPP | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| This antibody recognizes cytokeratin 10 (56.5kDa) and cytokeratin 13 (53kDa) in Western blotting. It recognizes only cytokeratin 13 in formalin-fixed, paraffin-embedded tissue sections. It does not react with cytokeratin 10 positive, cytokeratin 13 negative epithelia such as epidermis. However, on frozen sections this MAb serves as differentiation-related marker of all stratified epithelia; it stains all suprabasal cells in both cornifying and non-cornifying stratified epithelia and more differentiated cells of squamous carcinomas | |
| Store at 4C. | |
| IgG2a κ |
| Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin), SDS-Page, Immunofluorescence | |
| DE-K13 | |
| Unconjugated | |
| 10mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
| KRT10 | |
| Cytoskeletal preparation extracted from human ectocervical epithelium. | |
| 0.1 mg | |
| Cytoskeleton Markers | |
| 3858 | |
| Human, Feline | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction