missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Cytokeratin 1 Antibody (KRT1/8342R), Novus Biologicals™
Rabbit Monoclonal Antibody
£214.00 - £377.00
Specifications
| Antigen | Cytokeratin 1 |
|---|---|
| Clone | KRT1/8342R |
| Concentration | 0.2 mg/mL |
| Dilution | Immunohistochemistry-Paraffin 1-2 μg/mL for 30 minutes at RT |
| Applications | Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30043468
|
Novus Biologicals™
NBP3-26809-100ug |
100 μg |
£377.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30043244
|
Novus Biologicals™
NBP3-26809-20ug |
20 μg |
£214.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cytokeratin 1 Monoclonal antibody specifically detects Cytokeratin 1 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| Cytokeratin 1 | |
| 0.2 mg/mL | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Cellular Markers, Protein Kinase, Signal Transduction | |
| 10 mM PBS with 0.05% BSA | |
| 3848 | |
| IgG κ | |
| Protein A or G purified |
| KRT1/8342R | |
| Immunohistochemistry-Paraffin 1-2 μg/mL for 30 minutes at RT | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CK1, CK-1, cytokeratin 1, Cytokeratin-1, cytokeratin-1,67 kDa cytokeratin, EHK, EHK1, epidermolytic hyperkeratosis 1, EPPK, Hair alpha protein, K1, keratin 1, keratin, type II cytoskeletal 1, Keratin-1, KRT1A, KRTA, NEPPK, Type-II keratin Kb1 | |
| Recombinant full-length human Cytokeratin 1 protein | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title