missing translation for 'onlineSavingsMsg'
Learn More

Cytokeratin 1 Antibody (3C10F7), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 1 publication

Brand:  Novus Biologicals NB110-89450-0.025ml

 View more versions of this product

Product Code. 18596164

  • £144.00 / 25µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Cytokeratin 1 Monoclonal specifically detects Cytokeratin 1 in Human, Porcine, Bovine, Canine, Equine, Feline, Goat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry Free-Floating.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Cytokeratin 1
Monoclonal
Unconjugated
Ascites with 0.03% Sodium Azide
KRT1
Ni-NTA purified truncated recombinant CK1 expressed in E. Coli strain BL21 (DE3)
0.025 mL
Protein Kinase
3848
Human
Ascites
Western Blot
3C10F7
Western Blot 1:500 - 1:2000, ELISA 1:10000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 2-5 μg/mL, Immunohistochemistry-Paraffin 1:200-1:1000, Immunohistochemistry Free-Floating
CK1, CK-1, cytokeratin 1, Cytokeratin-1, cytokeratin-1,67 kDa cytokeratin, EHK, EHK1, epidermolytic hyperkeratosis 1, EPPK, Hair alpha protein, K1, keratin 1, keratin, type II cytoskeletal 1, Keratin-1, KRT1A, KRTA, NEPPK, Type-II keratin Kb1
Mouse
Unpurified
RUO
Primary
CK1 (PMC01)
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only