missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Cytohesin 4 Polyclonal antibody specifically detects Cytohesin 4 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA
Specifications
Specifications
| Antigen | Cytohesin 4 |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500, ELISA 1:10000 |
| Formulation | Contains Tris, HCl/glycine buffer (pH 7.4-7.8), 30% glycerol, 0.5% BSA, along with cryo-protective agents, and HEPES (exact storage and stabilization buffer information is proprietary) with 0.02% Sodium Azide |
| Gene Alias | CYT4PH, SEC7 and coiled-coil domain-containing protein 4, cytohesin 4, cytohesin-4, pleckstrin homology, Sec7 and coiled/coil domains 4, pleckstrin homology, Sec7 and coiled-coil domains 4, PSCD4DJ63G5.1 |
| Host Species | Rabbit |
| Immunogen | Synthetic peptide taken within amino acid region 300-362 on human Cytohesin 4 protein. |
| Purification Method | Affinity-purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?