missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytohesin 2 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB100-77340
This item is not returnable.
View return policy
Description
Cytohesin 2 Polyclonal specifically detects Cytohesin 2 in Human samples. It is validated for Western Blot, Peptide ELISA.
Specifications
| Cytohesin 2 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| ARF exchange factor, ARF nucleotide-binding site opener, ARNOPH, SEC7 and coiled-coil domain-containing protein 2, CTS18.1, cytohesin 2, pleckstrin homology, Sec7 and coiled-coil domains 2 (cytohesin-2), Protein ARNO, PSCD2LCTS18, PSCD2pleckstrin homology, Sec7 and coiled-coil domains 2, Sec7 and coiled/coil domains 2 (cytohesin-2) | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| This antibody is expected to recognize both reported isoforms (NP_004219.2; NP_059431.1). | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot, ELISA | |
| 0.5 mg/mL | |
| Western Blot 0.5 - 1.5 ug/ml, Peptide ELISA Detection limit 1:32000 | |
| NP_004219.2 | |
| CYTH2 | |
| Peptide with sequence EDGVYEPPDLTP-C corresponding to N-Terminus according to NP_059431.1. | |
| 0.1 mg | |
| Membrane Trafficking and Chaperones | |
| 9266 | |
| Human, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction