missing translation for 'onlineSavingsMsg'
Learn More

Cytohesin 2 Antibody (10A12), Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-22514-0.025mg

 View more versions of this product

Product Code. 18773114

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Cytohesin 2 Monoclonal specifically detects Cytohesin 2 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Western Blot (Negative).
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Cytohesin 2
Monoclonal
1 mg/mL
Flow Cytometry 3 - 5 μg/ 106 cells, Immunohistochemistry 1:50, Immunocytochemistry/Immunofluorescence 1:20, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:25 - 1:200, Western Blot (Negative)
ARF exchange factor, ARF nucleotide-binding site opener, ARNOPH, SEC7 and coiled-coil domain-containing protein 2, CTS18.1, cytohesin 2, pleckstrin homology, Sec7 and coiled-coil domains 2 (cytohesin-2), Protein ARNO, PSCD2LCTS18, PSCD2pleckstrin homology, Sec7 and coiled-coil domains 2, Sec7 and coiled/coil domains 2 (cytohesin-2)
Mouse
PEG purified
RUO
Primary
This antibody reacts with Human.
Store at -20°C. Avoid freeze-thaw cycles.
IgG1
Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
10A12
Unconjugated
Q99418
CYTH2
Purified, His-tagged cytohesin-2.
0.025 mg
Membrane Trafficking and Chaperones
9266
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only