missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cytohesin 1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA527793
This item is not returnable.
View return policy
Description
Recommended positive controls: PSCD1 tansfected 293T cell. Predicted reactivity: Mouse (97%), Rat (97%), Chicken (97%), Rhesus Monkey (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene is a member of the protein tyrosine phosphatase (PTP) family. PTPs are known to be signaling molecules that regulate a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. This PTP possesses an extracellular region, a single transmembrane region, and a single intracellular catalytic domains, and thus represents a receptor-type PTP. The similar gene predominately expressed in mouse brain was found to associate with, and thus regulate the activity and cellular localization of MAP kinases. The rat counterpart of this gene was reported to be regulated by the nerve growth factor, which suggested the function of this gene in neuronal growth and differentiation.
Specifications
| Cytohesin 1 | |
| Polyclonal | |
| Unconjugated | |
| CYTH1 | |
| B2-1; CLM1; CTH-1; CYTH1; CYTIP; cytoadhesin 1; cytohesin 1; CYTOHESIN-1; D17S811E; FLJ34050; FLJ41900; homolog of secretory protein SEC7; mKIAA4240; mSec7-1; PH, SEC7 and coiled-coil domain-containing protein 1; pleckstrin homology, Sec7 and coiled/coil domains 1(cytohesin 1); pleckstrin homology, Sec7 and coiled-coil domains 1; Pscd1; rSec7-1; SEC7; SEC7 homolog A; SEC7 homolog B2-1; Sec7a | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9267 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.87 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| Q15438 | |
| CYTH1 | |
| Recombinant protein encompassing a sequence within the center region of human Cytohesin 1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction