missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cytoglobin Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA575671
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
Cytoglobin is a ubiquitously expressed hexacoordinate hemoglobin that may facilitate diffusion of oxygen through tissues, scavenge nitric oxide or other reactive oxygen species, or serve a protective function during oxidative stress (Trent and Hargrove, 2002).
Specifications
| Cytoglobin | |
| Polyclonal | |
| Unconjugated | |
| CYGB | |
| 3110001K20Rik; Cygb; Cytoglobin; HGb; Histoglobin; Staap; STAP; stellate cell activation associated protein; stellate cell activation-associated protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 114757, 114886, 170520 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8WWM9, Q921A4, Q9CX80 | |
| CYGB | |
| Synthetic peptide corresponding to residues in Human Cytoglobin. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction