missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Cytodex 1 Gamma Microcarriers

Product Code. 15737207 Shop All Cytiva Products
Click to view available options
Quantity:
30 g
300 g
3 kg
Unit Size:
300g
30g
3kg
This item is not returnable. View return policy

Product Code. 15737207

Brand: Cytiva 17548703

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Microcarrier for general use in sterilized dry powder form.

  • Cytodex 1 Gamma is designed by substituting a cross-linked dextran matrix with positively charged N, N-diethylaminoethyl (DEAE) groups distributed throughout the matrix.
  • Micro-Carriers Dry Weight: 4.3 × 106
  • Surface Area Dry Weight: 0.44 m2/g
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Applications include the production of large quantities of cells, viruses and recombinant cell products (e.g. interferon, enzymes, nucleic acids, hormones), studies on cell adhesion, diff erentiation and cell function, perfusion column culture systems, microscopy studies, harvesting mitotic cells, isolation of cells, membrane studies, storage and transport of cells, assays involving celltransfer and studies on uptage of labelled compounds
Quantity 3 kg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.