missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Cytochrome P450 Pan Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA5101312

Product Code. 16617794

  • £458.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Antibody detects endogenous levels of total Cytochrome P450 2C8/9/18/19.

CYP2C9 is a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. CYP2C9 localizes to the endoplasmic reticulum and its expression is induced by rifampin. The enzyme is known to metabolize many xenobiotics, including phenytoin, tolbutamide, ibuprofen and S-warfarin.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Cytochrome P450 Pan
Polyclonal
Unconjugated
CYP2C9
(R)-limonene 6-monooxygenase; (S)-limonene 6-monooxygenase; (S)-limonene 7-monooxygenase; (S)-mephenytoin hydroxylase associated cytochrome P450; cholesterol 25-hydroxylase; CPC8; CPC9; CPCI; CPCJ; CYP2C; CYP2C10; CYP2C17; CYP2C18; CYP2C19; CYP2C8; CYP2C9; CYPIIC17; CYPIIC18; CYPIIC19; CYPIIC8; CYPIIC9; Cytochrome P450 2C18; cytochrome P450 2C19; Cytochrome P450 2C8; cytochrome P450 2C9; cytochrome P450 family 2 subfamily C member 18; cytochrome P450 family 2 subfamily C member 19; cytochrome P450 family 2 subfamily C member 8; cytochrome P450 family 2 subfamily C member 9; cytochrome P450 form 1; cytochrome P-450 II C; Cytochrome P450 IIC2; cytochrome P450 MP-12; Cytochrome P450 MP-20; Cytochrome P450 MP-4; Cytochrome P450 MP-8; Cytochrome P450 PB-1; cytochrome P-450 S-mephenytoin 4-hydroxylase; cytochrome P450, family 2, subfamily C, polypeptide 18; cytochrome P450, family 2, subfamily C, polypeptide 19; cytochrome P450, family 2, subfamily C, polypeptide 8; cytochrome P450, family 2, subfamily C, polypeptide 9; cytochrome P450, subfamily IIC (mephenytoin 4-hydroxylase), polypeptide 17; cytochrome P450, subfamily IIC (mephenytoin 4-hydroxylase), polypeptide 18; cytochrome P450, subfamily IIC (mephenytoin 4-hydroxylase), polypeptide 19; cytochrome P450, subfamily IIC (mephenytoin 4-hydroxylase), polypeptide 8; cytochrome P450-11A; Cytochrome P450-254C; cytochrome P450-6B/29C; cytochrome P-450MP; Fenbendazole monooxygenase (4'-hydroxylating); flavoprotein-linked monooxygenase; mephenytoin 4'-hydroxylase; mephenytoin 4-hydroxylase; microsomal monooxygenase; MP-12/MP-20; P450 form 1; P450-6B/29C; P450C2C; P450IIC17; P450IIC19; P450IIC9; s-mephenytoin 4-hydroxylase; unspecific monooxygenase; xenobiotic monooxygenase
Rabbit
Affinity Chromatography
RUO
13106, 1557, 1558, 1559, 1562, 1571
-20°C
Liquid
Western Blot, Immunocytochemistry
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P05181, P10632, P11712, P33260, P33261, Q05421
CYP2C18, CYP2C19, CYP2C8, CYP2C9, CYP2E1
A synthesized peptide derived from human CYP2C18(Accession P33260), corresponding to amino acid residues N116-P166.
100 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.