missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytochrome P450 4F11 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-16082
This item is not returnable.
View return policy
Description
Cytochrome P450 4F11 Polyclonal specifically detects Cytochrome P450 4F11 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Cytochrome P450 4F11 | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
| CYPIVF11, cytochrome P450 4F11, cytochrome P450, family 4, subfamily F, polypeptide 11, cytochrome P450, subfamily IVF, polypeptide 11, EC 1.14.14.1 | |
| Rabbit | |
| 60 kDa | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1mg/mL | |
| Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
| Q9HBI6 | |
| CYP4F11 | |
| Recombinant protein encompassing a sequence within the center region of human CYP4F11. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 57834 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction