missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytochrome P450 4A2, 4A3 Antibody (Clo4), Alexa Fluor™ 700, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Cytochrome P450 4A2, 4A3 Monoclonal specifically detects Cytochrome P450 4A2, 4A3 in Rat samples. It is validated for Western Blot, ELISA.
Specifications
Specifications
| Antigen | Cytochrome P450 4A2, 4A3 |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Clone | Clo4 |
| Conjugate | Alexa Fluor 700 |
| Dilution | Western Blot, ELISA |
| Formulation | 50mM Sodium Borate with 0.05% Sodium Azide |
| Host Species | Mouse |
| Immunogen | Rat liver cytochrome P450 4A2 and 4A3 |
| Purification Method | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?