missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytochrome P450 4A1, 2C11 Antibody (Clo1), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50211AF594
This item is not returnable.
View return policy
Description
Cytochrome P450 4A1, 2C11 Monoclonal specifically detects Cytochrome P450 4A1, 2C11 in Rat samples. It is validated for Western Blot, ELISA.
Specifications
| Cytochrome P450 4A1, 2C11 | |
| Monoclonal | |
| Alexa Fluor 594 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| Mouse | |
| Protein A purified | |
| Cancer, Cardiovascular Biology, Cellular Signaling, metabolism | |
| Rat | |
| IgG2b |
| Western Blot, ELISA | |
| Clo1 | |
| Western Blot, ELISA | |
| CYP4A1 | |
| Rat liver cytochrome P450 2C11 and 4A1 | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction