missing translation for 'onlineSavingsMsg'
Learn More

Cytochrome P450 4A Rabbit anti-Human, Mouse, Rat, Clone: 10D10W7, Novus Biologicals™

Product Code. 18359744 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18359744 20 μg 20µL
18350403 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18359744 Supplier Novus Biologicals Supplier No. NBP31584720UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Cytochrome P450 4A Monoclonal antibody specifically detects Cytochrome P450 4A in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytochrome P450 4A
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 10D10W7
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias 20-HETE synthase, CP4Y, CYP4A2, cytochrome P450 4A11, cytochrome P450, family 4, subfamily A, polypeptide 11, cytochrome P450, subfamily IVA, polypeptide 11, cytochrome P-450HK-omega, cytochrome P450HL-omega, fatty acid omega-hydroxylase, lauric acid omega-hydroxylase, P450HL-omega
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 50-150 of human Cytochrome P450 4A (CYP4A11) (Q02928).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 1579
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.