missing translation for 'onlineSavingsMsg'
Learn More

Cytochrome P450 4A Antibody (M25-P2A10), CoraFluor™ 1, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-50212CL1

Product Code. 30150652

  • £680.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Cytochrome P450 4A Monoclonal antibody specifically detects Cytochrome P450 4A in Drosophila,Human,Mouse,Rat,Xenopus samples. It is validated for ELISA,Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Cytochrome P450 4A
Monoclonal
CoraFluor 1
20-HETE synthase, CP4Y, CYP4A2, cytochrome P450 4A11, cytochrome P450, family 4, subfamily A, polypeptide 11, cytochrome P450, subfamily IVA, polypeptide 11, cytochrome P-450HK-omega, cytochrome P450HL-omega, fatty acid omega-hydroxylase, lauric acid omega-hydroxylase, P450HL-omega
Peptide Sequence - KNGIHLRLR (amino acids 499 - 507)
0.1 mL
Lipid and Metabolism
1579
Store at 4°C in the dark. Do not freeze.
IgG1 κ
ELISA, Immunohistochemistry, Western Blot
M25-P2A10
PBS
Mouse
Protein A purified
RUO
Primary
Drosophila, Human, Mouse, Rat, Xenopus species
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.