missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytochrome P450 3A4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NB600-1396-0.025ml
This item is not returnable.
View return policy
Description
Cytochrome P450 3A4 Polyclonal specifically detects Cytochrome P450 3A4 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| Cytochrome P450 3A4 | |
| Polyclonal | |
| Western Blot 1:1000 - 1:5000 | |
| Albendazole monooxygenase, Albendazole sulfoxidase, CP33, CP34, CYP3A, CYP3A3, CYPIIIA3, CYPIIIA4, Cytochrome P450 3A3, cytochrome P450 3A4, Cytochrome P450 HLp, Cytochrome P450 NF-25, cytochrome P450, family 3, subfamily A, polypeptide 4, cytochrome P450, subfamily IIIA (niphedipine oxidase), polypeptide 3, cytochrome P450, subfamily IIIA (niphedipine oxidase), polypeptide 4, Cytochrome P450-PCN1, EC 1.14.13.32, EC 1.14.13.67, EC 1.14.13.97, EC 1.14.14.1, glucocorticoid-inducible P450, HLP, MGC126680, NF-25, Nifedipine oxidase, P450C3, P450-III, steroid inducible, P450PCN1, Quinine 3-monooxygenase, Taurochenodeoxycholate 6-alpha-hydroxylase | |
| Rabbit | |
| Ammonium sulfate precipitation | |
| RUO | |
| Primary | |
| Detects Cytochrome P450 (P450) 3A4 from human and rat tissues and cells. This detects, to a lesser extent, other P450 isoforms. | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| CYP3A4 | |
| Purified, His-tagged, full length human P450 3A4 fusion protein. | |
| 0.025 mL | |
| Lipid and Metabolism | |
| 1576 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction