missing translation for 'onlineSavingsMsg'
Learn More

Cytochrome P450 3A4 Antibody, DyLight 405, Novus Biologicals™

Product Code. 18377973 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18377973 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18377973 Supplier Novus Biologicals Supplier No. NBP299089V

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Cytochrome P450 3A4 Polyclonal specifically detects Cytochrome P450 3A4 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytochrome P450 3A4
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 405
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias Albendazole monooxygenase, Albendazole sulfoxidase, CP33, CP34, CYP3A, CYP3A3, CYPIIIA3, CYPIIIA4, Cytochrome P450 3A3, cytochrome P450 3A4, Cytochrome P450 HLp, Cytochrome P450 NF-25, cytochrome P450, family 3, subfamily A, polypeptide 4, cytochrome P450, subfamily IIIA (niphedipine oxidase), polypeptide 3, cytochrome P450, subfamily IIIA (niphedipine oxidase), polypeptide 4, Cytochrome P450-PCN1, EC 1.14.13.32, EC 1.14.13.67, EC 1.14.13.97, EC 1.14.14.1, glucocorticoid-inducible P450, HLP, MGC126680, NF-25, Nifedipine oxidase, P450C3, P450-III, steroid inducible, P450PCN1, Quinine 3-monooxygenase, Taurochenodeoxycholate 6-alpha-hydroxylase
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human Cytochrome P450 3A4.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 1576
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.