missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Cytochrome P450 3A4 Polyclonal specifically detects Cytochrome P450 3A4 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
| Antigen | Cytochrome P450 3A4 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 647 |
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin |
| Formulation | 50mM Sodium Borate |
| Gene Alias | Albendazole monooxygenase, Albendazole sulfoxidase, CP33, CP34, CYP3A, CYP3A3, CYPIIIA3, CYPIIIA4, Cytochrome P450 3A3, cytochrome P450 3A4, Cytochrome P450 HLp, Cytochrome P450 NF-25, cytochrome P450, family 3, subfamily A, polypeptide 4, cytochrome P450, subfamily IIIA (niphedipine oxidase), polypeptide 3, cytochrome P450, subfamily IIIA (niphedipine oxidase), polypeptide 4, Cytochrome P450-PCN1, EC 1.14.13.32, EC 1.14.13.67, EC 1.14.13.97, EC 1.14.14.1, glucocorticoid-inducible P450, HLP, MGC126680, NF-25, Nifedipine oxidase, P450C3, P450-III, steroid inducible, P450PCN1, Quinine 3-monooxygenase, Taurochenodeoxycholate 6-alpha-hydroxylase |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human Cytochrome P450 3A4. |
| Purification Method | Antigen Affinity-purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?