missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytochrome p450 2J2 Antibody (OTI5C5), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70539PE
This item is not returnable.
View return policy
Description
Cytochrome p450 2J2 Monoclonal antibody specifically detects Cytochrome p450 2J2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry
Specifications
| Cytochrome p450 2J2 | |
| Monoclonal | |
| PE | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| OTI5C5 | |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| Arachidonic acid epoxygenase, CPJ2, CYPIIJ2, cytochrome P450 2J2, cytochrome P450, family 2, subfamily J, polypeptide 2, cytochrome P450, subfamily IIJ (arachidonic acid epoxygenase) polypeptide 2, EC 1.14.14.1, flavoprotein-linked monooxygenase, microsomal monooxygenase | |
| Full length human recombinant protein of human CYP2J2(NP_000766) produced in HEK293T cell. | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 1573 | |
| Store at 4C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction