missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytochrome P450 2E1 Mouse anti-Human, Clone: M12P4H2, Novus Biologicals™
Mouse Monoclonal Antibody
£396.00
Specifications
| Antigen | Cytochrome P450 2E1 |
|---|---|
| Clone | M12P4H2 |
| Concentration | 0.2 mg/mL |
| Dilution | Immunohistochemistry-Paraffin 0.1-0.2 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
Description
Cytochrome P450 2E1 Monoclonal specifically detects Cytochrome P450 2E1 in Human samples. It is validated for Immunohistochemistry-Paraffin.Specifications
| Cytochrome P450 2E1 | |
| 0.2 mg/mL | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Lipid and Metabolism | |
| 10 mM PBS with 0.05% BSA | |
| 1571 | |
| IgG1 κ | |
| Protein A or G purified |
| M12P4H2 | |
| Immunohistochemistry-Paraffin 0.1-0.2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| 4-nitrophenol 2-hydroxylase, CPE1, CYP2Ecytochrome P450 2E1, CYPIIE1, cytochrome P450, family 2, subfamily E, polypeptide 1, cytochrome P450, subfamily IIE (ethanol-inducible), polypeptide 1, Cytochrome P450-J, EC 1.14.13.-, EC 1.14.13.n7, EC 1.14.14.1, flavoprotein-linked monooxygenase, microsomal monooxygenase, P450C2E, P450-J, xenobiotic monooxygenase | |
| Ovalbumin-conjugated synthetic peptide HIGFGCIPPR (C-terminal sequence) | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title