missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytochrome P450 2E1 Antibody (M12P4H2) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50203
This item is not returnable.
View return policy
Description
Cytochrome P450 2E1 Monoclonal antibody specifically detects Cytochrome P450 2E1 in Human samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| Cytochrome P450 2E1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500 | |
| 4-nitrophenol 2-hydroxylase, CPE1, CYP2Ecytochrome P450 2E1, CYPIIE1, cytochrome P450, family 2, subfamily E, polypeptide 1, cytochrome P450, subfamily IIE (ethanol-inducible), polypeptide 1, Cytochrome P450-J, EC 1.14.13.-, EC 1.14.13.n7, EC 1.14.14.1, flavoprotein-linked monooxygenase, microsomal monooxygenase, P450C2E, P450-J, xenobiotic monooxygenase | |
| Ovalbumin-conjugated synthetic peptide HIGFGCIPPR (C-terminal sequence) | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 1571 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 λ |
| Western Blot, Immunohistochemistry | |
| M12P4H2 | |
| Unconjugated | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction