missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytochrome P450 2D6 Rabbit anti-Human, Mouse, Rat, Clone: 1X3F0, Novus Biologicals™
Rabbit Monoclonal Antibody
£150.00 - £386.00
Specifications
| Antigen | Cytochrome P450 2D6 |
|---|---|
| Clone | 1X3F0 |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18389641
|
Novus Biologicals
NBP3-16610-20UL |
20 μg |
£150.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18334053
|
Bio-Techne
NBP3-16610-100UL |
100 μg |
£386.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cytochrome P450 2D6 Monoclonal antibody specifically detects Cytochrome P450 2D6 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, ImmunofluorescenceSpecifications
| Cytochrome P450 2D6 | |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cardiovascular Biology, Endocrinology, Signal Transduction | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 1565 | |
| IgG | |
| Affinity purified |
| 1X3F0 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| CPD6P450DB1, CYP2D, CYP2D7AP, CYP2D7BP, CYP2D7P2, CYP2D8P2, CYP2DL1, CYPIID6, cytochrome P450 2D6, cytochrome P450, family 2, subfamily D, polypeptide 6, cytochrome P450, family 2, subfamily D, polypeptide 7 pseudogene 2, cytochrome P450, family 2, subfamily D, polypeptide 8 pseudogene 2, cytochrome P450, subfamily II (debrisoquine, sparteine, etc., -metabolising), cytochrome P450, subfamily IID (debrisoquine, sparteine, etc., cytochrome P450, subfamily IID (debrisoquine, sparteine, etc., -metabolising), cytochrome P450, subfamily IID (debrisoquine, sparteine, etc., -metabolizing), Cytochrome P450-DB1, Debrisoquine 4-hydroxylase, EC 1.14.14.1, flavoprotein-linked monooxygenase, -metabolizing)-like 1, MGC120389, MGC120390, microsomal monooxygenase, P450C2D, P450-DB1, polypeptide 6, polypeptide 7 pseudogene 2, polypeptide 8 pseudogene 2, xenobiotic monooxygenase | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 341-497 of human Cytochrome P450 2D6 (P10635). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title