missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytochrome P450 2D6 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35296-100ul
This item is not returnable.
View return policy
Description
Cytochrome P450 2D6 Polyclonal antibody specifically detects Cytochrome P450 2D6 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| Cytochrome P450 2D6 | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000, ELISA | |
| CPD6P450DB1, CYP2D, CYP2D7AP, CYP2D7BP, CYP2D7P2, CYP2D8P2, CYP2DL1, CYPIID6, cytochrome P450 2D6, cytochrome P450, family 2, subfamily D, polypeptide 6, cytochrome P450, family 2, subfamily D, polypeptide 7 pseudogene 2, cytochrome P450, family 2, subfamily D, polypeptide 8 pseudogene 2, cytochrome P450, subfamily II (debrisoquine, sparteine, etc., -metabolising), cytochrome P450, subfamily IID (debrisoquine, sparteine, etc., cytochrome P450, subfamily IID (debrisoquine, sparteine, etc., -metabolising), cytochrome P450, subfamily IID (debrisoquine, sparteine, etc., -metabolizing), Cytochrome P450-DB1, Debrisoquine 4-hydroxylase, EC 1.14.14.1, flavoprotein-linked monooxygenase, -metabolizing)-like 1, MGC120389, MGC120390, microsomal monooxygenase, P450C2D, P450-DB1, polypeptide 6, polypeptide 7 pseudogene 2, polypeptide 8 pseudogene 2, xenobiotic monooxygenase | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 20-230 of human Cytochrome P450 2D6 (NP_000097.3).,, Sequence:, VDLMHRRQRWAARYPPGPLPLPGLGNLLHVDFQNTPYCFDQLRRRFGDVFSLQLAWTPVVVLNGLAAVREALVTHGEDTADRPPVPITQILGFGPRSQGVFLARYGPAWREQRRFSVSTLRNLGLGKKSLEQWVTEEAACLCAAFANHSGRPFRPNGLLDKAVSNVIASLTCGRRFEYDDPRFLRLLDLAQEGLKEESGFLREVLNAVPVL | |
| 100 μL | |
| Cancer, Cardiovascular Biology, Endocrinology, Signal Transduction | |
| 1565 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction