missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytochrome P450 2C9 Antibody (CYP2C9/13121), Novus Biologicals™
Mouse Monoclonal Antibody
£195.00 - £368.00
Specifications
| Antigen | Cytochrome P450 2C9 |
|---|---|
| Clone | CYP2C9/13121 |
| Concentration | 0.2 mg/ml |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30301361
|
Novus Biologicals
NBP3-44591-20ug |
20 μg |
£195.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30299390
|
Novus Biologicals
NBP3-44591-100ug |
100 μg |
£368.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cytochrome P450 2C9 Monoclonal antibody specifically detects Cytochrome P450 2C9 in Human samples. It is validated for nanSpecifications
| Cytochrome P450 2C9 | |
| 0.2 mg/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| (R)-limonene 6-monooxygenase, (S)-limonene 6-monooxygenase, (S)-limonene 7-monooxygenase, CPC9, CYP2C, CYP2C10, CYPIIC9, cytochrome P450 2C9, Cytochrome P450 MP-4, Cytochrome P450 MP-8, Cytochrome P450 PB-1, cytochrome P-450 S-mephenytoin 4-hydroxylase, cytochrome P450, family 2, subfamily C, polypeptide 9, cytochrome P450, subfamily IIC (mephenytoin 4-hydroxylase), polypeptide 9, Cytochrome P-450MP, EC 1.14.13.48, EC 1.14.13.49, EC 1.14.13.80, EC 1.14.14.1, flavoprotein-linked monooxygenase, MGC149605, MGC88320, microsomal monooxygenase, P450IIC9, S-mephenytoin 4-hydroxylase, xenobiotic monooxygenase | |
| Recombinant fragment (around aa1-200) of human CYP2C9 protein (Uniprot: P11712). Exact sequence is proprietary. | |
| Primary | |
| Store at 4⪚C. |
| CYP2C9/13121 | |
| Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Lipid and Metabolism | |
| 10 mM PBS with 0.05% BSA | |
| 1559 | |
| IgG2b κ | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title