missing translation for 'onlineSavingsMsg'
Learn More

Cytochrome P450 2B1, 2B2 Antibody (b/e3) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody

£150.00 - £302.00

Specifications

Antigen Cytochrome P450 2B1, 2B2
Clone b/e3
Concentration 0.84 mg/mL
Dilution Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunoprecipitation 1:10 - 1:500
Applications Western Blot, ELISA, Immunoprecipitation
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18645718
View Documents
Novus Biologicals
NBP2-50201-0.025ml
0.025 mL
£150.00
0.03mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18697246
View Documents
Novus Biologicals
NBP2-50201
0.1 mL
£302.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Cytochrome P450 2B1, 2B2 Monoclonal antibody specifically detects Cytochrome P450 2B1, 2B2 in Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunoprecipitation
Specifications

Specifications

Cytochrome P450 2B1, 2B2
0.84 mg/mL
Western Blot, ELISA, Immunoprecipitation
Unconjugated
Mouse
Cancer, Cardiovascular Biology, Cellular Signaling, metabolism
PBS
IgG1
Protein A purified
b/e3
Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunoprecipitation 1:10 - 1:500
Monoclonal
Purified
RUO
Mouse, Rat
Rat liver cytochromes p450 2B1 and 2B2
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.