missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Cytochrome P450 26B1 Antibody (1H6), Novus Biologicals™

Product Code. 18347839 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18347839 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18347839 Supplier Novus Biologicals™ Supplier No. H00056603M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Cytochrome P450 26B1 Monoclonal antibody specifically detects Cytochrome P450 26B1 in Human,Monkey samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytochrome P450 26B1
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1H6
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_063938
Gene Alias CYP26A2, Cytochrome P450 26A2, cytochrome P450 26B1, cytochrome P450 CYP26B1 variant 2, Cytochrome P450 retinoic acid-inactivating 2, cytochrome P450 retinoid metabolizing protein, cytochrome P450, family 26, subfamily B, polypeptide 1, Cytochrome P450RAI-2, DKFZp686G0638, EC 1.14, MGC129613, P450RAI2, P450RAI-2, Retinoic acid-metabolizing cytochrome
Host Species Mouse
Immunogen CYP26B1 (NP_063938, 131 a.a. ∽ 230 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. SNSIGDIHRNKRKVFSKIFSHEALESYLPKIQLVIQDTLRAWSSHPEAINVYQEAQKLTFRMAIRVLLGFSIPEEDLGHLFEVYQQFVDNVFSLPVDLPF
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cancer, Cardiovascular Biology, Cellular Signaling, metabolism
Primary or Secondary Primary
Gene ID (Entrez) 56603
Target Species Human, Monkey
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.