missing translation for 'onlineSavingsMsg'
Learn More

Cytochrome P450 1A2 Mouse anti-Human, Clone: F16 P2 D8, Novus Biologicals™

Product Code. 18326892 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
200 μg
25 μg
Unit Size:
200µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18326892 200 μg 200µg
18379501 25 μg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18326892 Supplier Novus Biologicals Supplier No. NBP311912200UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Cytochrome P450 1A2 Monoclonal antibody specifically detects Cytochrome P450 1A2 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytochrome P450 1A2
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone F16 P2 D8
Conjugate Unconjugated
Dilution Western Blot 1 - 2 ug/ml, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Formulation PBS
Gene Alias aryl hydrocarbon hydroxylase, CP12, CYPIA2, Cytochrome P(3)450, cytochrome P450 1A2, Cytochrome P450 4, cytochrome P450, family 1, subfamily A, polypeptide 2, cytochrome P450, subfamily I (aromatic compound-inducible), polypeptide 2, Cytochrome P450-P3, dioxin-inducible P3-450, EC 1.14.14.1, flavoprotein-linked monooxygenase, microsomal monooxygenase, P3-450, P450 form 4, P450(PA), xenobiotic monooxygenase
Host Species Mouse
Immunogen Hybridoma produced by the fusion of splenocytes from BALB/c mice immunized with Synthetic peptide conjugated to ovalbumin derived from the C-terminus of CYP2A6 and mouse myeloma Ag8563 cells. Antigen identical in CYP2A6, 2A7 and 2A13. (Uniprot: P05177)
Purification Method Protein A or G purified
Quantity 200 μg
Regulatory Status RUO
Research Discipline Cancer, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 1544
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.