missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytochrome P450 1A2 Mouse anti-Human, Clone: F16 P2 D8, Novus Biologicals™
Mouse Monoclonal Antibody
£198.00 - £401.00
Specifications
| Antigen | Cytochrome P450 1A2 |
|---|---|
| Clone | F16 P2 D8 |
| Dilution | Western Blot 1 - 2 ug/ml, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18379501
|
Novus Biologicals
NBP3-11912-25UG |
25 μg |
£198.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18326892
|
Novus Biologicals
NBP3-11912-200UG |
200 μg |
£401.00
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cytochrome P450 1A2 Monoclonal antibody specifically detects Cytochrome P450 1A2 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)Specifications
| Cytochrome P450 1A2 | |
| Western Blot 1 - 2 ug/ml, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| aryl hydrocarbon hydroxylase, CP12, CYPIA2, Cytochrome P(3)450, cytochrome P450 1A2, Cytochrome P450 4, cytochrome P450, family 1, subfamily A, polypeptide 2, cytochrome P450, subfamily I (aromatic compound-inducible), polypeptide 2, Cytochrome P450-P3, dioxin-inducible P3-450, EC 1.14.14.1, flavoprotein-linked monooxygenase, microsomal monooxygenase, P3-450, P450 form 4, P450(PA), xenobiotic monooxygenase | |
| Hybridoma produced by the fusion of splenocytes from BALB/c mice immunized with Synthetic peptide conjugated to ovalbumin derived from the C-terminus of CYP2A6 and mouse myeloma Ag8563 cells. Antigen identical in CYP2A6, 2A7 and 2A13. (Uniprot: P05177) | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. |
| F16 P2 D8 | |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| Unconjugated | |
| Mouse | |
| Cancer, Lipid and Metabolism | |
| PBS | |
| 1544 | |
| IgG1 | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title