missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytochrome P450 1A2 Antibody (D15), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB600-1490B
This item is not returnable.
View return policy
Description
Cytochrome P450 1A2 Monoclonal specifically detects Cytochrome P450 1A2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.
Specifications
| Cytochrome P450 1A2 | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| CYP1A2 | |
| MC1a (Preparation C31B4, rat liver cytochrome P4501A2) | |
| 0.1 mL | |
| Cancer, Lipid and Metabolism | |
| 1544 | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| D15 | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| aryl hydrocarbon hydroxylase, CP12, CYPIA2, Cytochrome P(3)450, cytochrome P450 1A2, Cytochrome P450 4, cytochrome P450, family 1, subfamily A, polypeptide 2, cytochrome P450, subfamily I (aromatic compound-inducible), polypeptide 2, Cytochrome P450-P3, dioxin-inducible P3-450, EC 1.14.14.1, flavoprotein-linked monooxygenase, microsomal monooxygenase, P3-450, P450 form 4, P450(PA), xenobiotic monooxygenase | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| D15 cross reacts with CYP1A1 & CYP1A2. | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction