missing translation for 'onlineSavingsMsg'
Learn More

Cytochrome P450 1A1/1A2 Antibody (MC1), Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NB100-74398

Product Code. 18290721

  • £409.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Cytochrome P450 1A1/1A2 Monoclonal specifically detects Cytochrome P450 1A1/1A2 in Human, Mouse, Rat, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Cytochrome P450 1A1/1A2
Monoclonal
Unconjugated
Ascites with 0.05% Sodium Azide
CYP1A1
3-methylcholanthrene induced rat cytochrome P450 protein.
Unpurified
RUO
Primary
Detects Cytochrome P450 1A1 (Gene ID: 1543, UniProt P04798) and Cytochrom P450 1A2 (Gene ID: 1544, UniProt: P05177) from human samples.
Store at -20C. Avoid freeze-thaw cycles.
IgG1
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
MC1
Western Blot 1:500, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 1:20 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200
AHH, AHRR, aryl hydrocarbon hydroxylase, CP11, CYP1, CYPIA1, cytochrome P1-450, dioxin-inducible, cytochrome P450 1A1, Cytochrome P450 form 6, cytochrome P450, family 1, subfamily A, polypeptide 1, Cytochrome P450-C, Cytochrome P450-P1, EC 1.14.14.1, flavoprotein-linked monooxygenase, P1-450, P450-C, P450DX, subfamily I (aromatic compound-inducible), polypeptide 1, xenobiotic monooxygenase
Mouse
58 kDa
100 μL
Breast Cancer, Lipid and Metabolism
1543
Human, Mouse, Rat, Primate
Ascites
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only