missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytochrome P450 17A1 Rabbit anti-Human, Mouse, Clone: 5D2H1, Novus Biologicals™
Rabbit Monoclonal Antibody
£150.00 - £386.00
Specifications
| Antigen | Cytochrome P450 17A1 |
|---|---|
| Clone | 5D2H1 |
| Applications | Western Blot, Immunofluorescence |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18377836
|
Novus Biologicals
NBP3-16541-20UL |
20 μg |
£150.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18375822
|
Bio-Techne
NBP3-16541-100UL |
100 μg |
£386.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beskrivelse
Cytochrome P450 17A1 Monoclonal antibody specifically detects Cytochrome P450 17A1 in Human, Mouse samples. It is validated for Western Blot, ImmunofluorescenceTekniske data
| Cytochrome P450 17A1 | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Lipid and Metabolism | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 1586 | |
| IgG | |
| Affinity purified |
| 5D2H1 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| CPT7, cyp17, CYP17A1, cytochrome p450 XVIIA1, cytochrome P450, family 17, subfamily A, polypeptide 1, cytochrome P450, subfamily XVII (steroid 17-alpha-hydroxylase), adrenalhyperplasia, Cytochrome P450c17, Cytochrome P450-C17, EC 1.14.99, P450C17, P450C17CYPXVII, S17AH | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 101-220 of human Cytochrome P450 17A1 (P05093). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Ser du en mulighed for forbedring?Del en indholdskorrektion
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel