missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytochrome P450 17A1 Antibody (OTI3F12), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70542B
This item is not returnable.
View return policy
Description
Cytochrome P450 17A1 Monoclonal specifically detects Cytochrome P450 17A1 in Human samples. It is validated for Western Blot.
Specifications
| Cytochrome P450 17A1 | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| CYP17A1 | |
| Full length recombinant protein of human Cytochrome P450 17A1 (NP_000093) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot | |
| OTI3F12 | |
| Western Blot | |
| CPT7, cyp17, CYP17A1, cytochrome p450 XVIIA1, cytochrome P450, family 17, subfamily A, polypeptide 1, cytochrome P450, subfamily XVII (steroid 17-alpha-hydroxylase), adrenalhyperplasia, Cytochrome P450c17, Cytochrome P450-C17, EC 1.14.99, P450C17, P450C17CYPXVII, S17AH | |
| Mouse | |
| Affinity Purified | |
| Lipid and Metabolism | |
| 1586.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction