missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytochrome P450 17A1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35404-100ul
This item is not returnable.
View return policy
Description
Cytochrome P450 17A1 Polyclonal antibody specifically detects Cytochrome P450 17A1 in Rat samples. It is validated for ELISA,Western Blot
Specifications
| Cytochrome P450 17A1 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| CPT7, cyp17, CYP17A1, cytochrome p450 XVIIA1, cytochrome P450, family 17, subfamily A, polypeptide 1, cytochrome P450, subfamily XVII (steroid 17-alpha-hydroxylase), adrenalhyperplasia, Cytochrome P450c17, Cytochrome P450-C17, EC 1.14.99, P450C17, P450C17CYPXVII, S17AH | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 209-508 of human Cytochrome P450 17A1 (NP_000093.1).,, Sequence:, LSKDSLVDLVPWLKIFPNKTLEKLKSHVKIRNDLLNKILENYKEKFRSDSITNMLDTLMQAKMNSDNGNAGPDQDSELLSDNHILTTIGDIFGAGVETTTSVVKWTLAFLLHNPQVKKKLYEEIDQNVGFSRTPTISDRNRLLLLEATIREVLRLRPVAPMLIPHKANVDSSIGEFAVDKGTEVIINLWALHHNEKEWHQPDQFMPERFLNPAGTQLISPSVSYLPFGAGPRSCIGEILARQELFLIMAWLLQRFDLEVPDDGQLPSLEGIPKVVFLIDSFKVKIKVRQAWREAQAEGST | |
| 100 μL | |
| Lipid and Metabolism | |
| 1586 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction