missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cytochrome C Polyclonal Antibody

Description
A suggested positive control for this product is rat1 cell lysate.
Cytochrome C is an electron transporting protein that resides within the intermembrane space of the mitochondria, where it plays a critical role in oxidative phosphorylation and the production of cellular ATP. Release of cytochrome C from the mitochondria to the cytosol is triggered by apoptotic stimuli. In the cytoplasm, the cytochrome C binds apoptotic protease activating factor which activates the apoptotic initiator procaspase 9. Cytosolic cytochrome C functions in the activation of caspase 3, an ICE family molecule that is a key effector of apoptosis.
Specifications
Specifications
| Antigen | Cytochrome C |
| Applications | Immunohistochemistry, Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Clone | N/A |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.08% sodium azide |
| Gene | Cycs |
| Gene Accession No. | P00011, P62898, P99999 |
| Gene Alias | CG17903; CG17903-PA; CG17903-PB; CRF; CYC; CYC2; Cycs; CYCSA; Cyct-c-p; Cyt c; cyt c-p; cyt.c; CytC; Cyt-c; CYTC2; Cyt-c2; CytC-2; cyt-c-p; cytC-p; Cyt-c-p-PA; Cyt-c-p-PB; Cytochrome c; cytochrome c DC4; Cytochrome c proximal; Cytochrome C, expressed in somatic tissues; cytochrome c, somatic; Cytochrome c-2; cytochrome c-like; cytochrome c-like protein; Cytochrome c-proximal; cytochrome-c-p; Cytochrome-c-proximal; DC4; dCyt.C-p; DMc01; Dmel\CG17903; Dmel_CG17903; hc1; HCS; l(2)k13905; lethal (2) k13905; LOC100341175; LOC100357840; MGC128356 protein; pDMc01; RH17228p; THC4 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?