missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cytochrome C Monoclonal Antibody (7H8.2C12)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA511674
This item is not returnable.
View return policy
Description
MA5-11674 targets Cytochrome c in IHC (P) and WB applications and shows reactivity with Human, mouse, and Rat samples. The MA5-11674 immunogen is synthetic peptides corresponding to amino acids 1-80, 81-104 and 66-104 of pigeon cytochrome c.
Cytochrome C is an electron transporting protein that resides within the intermembrane space of the mitochondria, where it plays a critical role in oxidative phosphorylation and the production of cellular ATP. Release of cytochrome C from the mitochondria to the cytosol is triggered by apoptotic stimuli. In the cytoplasm, the cytochrome C binds apoptotic protease activating factor which activates the apoptotic initiator procaspase 9. Cytosolic cytochrome C functions in the activation of caspase 3, an ICE family molecule that is a key effector of apoptosis.
Specifications
| Cytochrome C | |
| Monoclonal | |
| 0.2 mg/mL | |
| PBS with 0.05% BSA and 0.05% sodium azide; pH 7.4 | |
| P62897, P99999 | |
| Cycs | |
| Synthetic peptides corresponding to amino acids 1-80, 81-104 and 66-104 of pigeon cytochrome c. | |
| 500 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG2b κ |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 7H8.2C12 | |
| Unconjugated | |
| Cycs | |
| CG17903; CG17903-PA; CG17903-PB; CRF; CYC; CYC2; Cycs; CYCSA; Cyct-c-p; Cyt c; cyt c-p; cyt.c; CytC; Cyt-c; CYTC2; Cyt-c2; CytC-2; cyt-c-p; cytC-p; Cyt-c-p-PA; Cyt-c-p-PB; Cytochrome c; cytochrome c DC4; Cytochrome c proximal; Cytochrome C, expressed in somatic tissues; cytochrome c, somatic; Cytochrome c-2; cytochrome c-like; cytochrome c-like protein; Cytochrome c-proximal; cytochrome-c-p; Cytochrome-c-proximal; DC4; dCyt.C-p; DMc01; Dmel\CG17903; Dmel_CG17903; hc1; HCS; l(2)k13905; lethal (2) k13905; LOC100341175; LOC100357840; MGC128356 protein; pDMc01; RH17228p; THC4 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 13063, 54205 | |
| 4°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction