missing translation for 'onlineSavingsMsg'
Learn More

Cytochrome c Antibody (CYCS/3128R), PerCP, R&D Systems™

Product Code. 18613958 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18613958 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18613958 Supplier Novus Biologicals Supplier No. NBP308598PCP
  • ONLINE EXCLUSIVE PRICE
    £564.48
    £481.95 / 100µL
    Save £82.53
    14% Off
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Rabbit Monoclonal Antibody

Cytochrome c Monoclonal specifically detects Cytochrome c in Amphibian/Canine/Drosophila/Equine/Human/Mouse/Pigeon/Rat samples. It is validated for Flow Cytometry/Immunocytochemistry/Immunofluorescence/Immunohistochemistry/Immunohistochemistry (Paraffin)/Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytochrome c
Applications Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone CYCS/3128R
Conjugate PerCP
Formulation PBS
Gene Alias CYCHCS, cytochrome c, cytochrome c, somatic, THC4
Host Species Rabbit
Immunogen Recombinant full-length human Cytochrome c protein (Uniprot: P99999)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis, Cellular Markers, Cholesterol Metabolism, Core ESC Like Genes, Lipid and Metabolism, Mitochondrial Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 54205
Target Species Amphibian, Canine, Drosophila, Equine, Human, Mouse, Pigeon, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.