Learn More
Invitrogen™ Cytochrome C Antibody Cocktail

Description
ApoTrack™ Cytochrome c WB Cocktail (Sku# 459160) provides a means to detect apoptosis while detecting release of cytochrome c, as well as a cytoplasmic and mitochondrial markers with the help of western blot. This cocktail provides a means to detect cytochrome c, as well as a cytoplasmic and mitochondrial markers Antibodies Cocktail Contains: Mouse anti Cyt. c monoclonal: Amount: 50 μg Working concentration: 1 μg/mL Mouse anti GAPDH monclonal: Amount: 5 μg Working concentration: 0.1 μg/mL Mouse anti PDH-E1-alpha monoclonal: Amount: 100 μg Working concentration: 2 μg/mL Mouse anti C-V-alpha monoclonal: Amount: 25 μg Working concentration: 0.5 μg/mL.
Specifications
Specifications
| Antigen | Cytochrome C |
| Applications | Western Blot |
| Classification | Cocktail |
| Clone | Cocktail |
| Concentration | 0.9 mg/mL |
| Conjugate | Unconjugated |
| Formulation | HEPES buffered saline with 0.02% sodium azide |
| Gene | Cycs |
| Gene Accession No. | P99999 |
| Gene Alias | CG17903; CG17903-PA; CG17903-PB; CRF; CYC; CYC2; Cycs; CYCSA; Cyct-c-p; Cyt c; cyt c-p; cyt.c; CytC; Cyt-c; CYTC2; Cyt-c2; CytC-2; cyt-c-p; cytC-p; Cyt-c-p-PA; Cyt-c-p-PB; Cytochrome c; cytochrome c DC4; Cytochrome c proximal; Cytochrome C, expressed in somatic tissues; cytochrome c, somatic; Cytochrome c-2; cytochrome c-like; cytochrome c-like protein; Cytochrome c-proximal; cytochrome-c-p; Cytochrome-c-proximal; DC4; dCyt.C-p; DMc01; Dmel\CG17903; Dmel_CG17903; hc1; HCS; l(2)k13905; lethal (2) k13905; LOC100341175; LOC100357840; MGC128356 protein; pDMc01; RH17228p; THC4 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.