missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytochrome c Antibody (2B5F8) [DyLight 680], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB898Y
This item is not returnable.
View return policy
Description
Cytochrome c Monoclonal antibody specifically detects Cytochrome c in Equine,Human samples. It is validated for Immunohistochemistry,Immunoprecipitation,Immunocytochemistry
Specifications
| Cytochrome c | |
| Monoclonal | |
| DyLight 680 | |
| CYCHCS, cytochrome c, cytochrome c, somatic, THC4 | |
| Equine Cytochrome c | |
| 0.1 mL | |
| Apoptosis, Cellular Markers, Cholesterol Metabolism, Core ESC Like Genes, Lipid and Metabolism, Mitochondrial Markers, Stem Cell Markers | |
| 54205 | |
| Store at 4°C in the dark. | |
| IgG2a |
| Immunohistochemistry, Immunoprecipitation, Immunocytochemistry | |
| 2B5F8 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| Primary | |
| Equine, Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction