missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cytochrome B5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA563584
This item is not returnable.
View return policy
Description
Immunogen sequence: MAEQSDEAVK YYTLEEIQKH NHSKSTWLIL HHKVYDLTKF LAEHPGG Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 87%.
Cytochrome b5 is a membrane-bound member of the cytochrome b family. A heme protein that functions as an electron carrier for many membrane-bound oxygenases, cytochrome b5 possesses two heme groups, which are not covalently attached to the protein. Two isoforms of cytochrome b5, a microsomal membrane-bound form and a cytoplasmic form, are produced by alternative splicing. Mutations in cytochrome b5 are associated with Leber's hereditary optic neuropathy and with myopathy.
Specifications
| Cytochrome B5 | |
| Polyclonal | |
| Unconjugated | |
| CYB5A | |
| 0610009N12Rik; Cyb5; CYB5A; cytochrome b5; cytochrome b-5; cytochrome b5 type A; cytochrome b5 type A (microsomal); MCB5; Microsomal cytochrome b5 type A; type 1 cyt-b5 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1528 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P00167 | |
| CYB5A | |
| Recombinant protein corresponding to Human Cytochrome B5. Recombinant protein control fragment (Product #RP-103776). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction